Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tcfl5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178254.3 | NP_839985.2 | 4 | 1149 | ||
GenBank mRNA | AK132721.1 | - | 4 | 809 | ||
AY234363.1 | - | 4 | 943 | |||
BC108399.1 | - | 4 | 1149 | 6394526 |