Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prex1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177782.3 | NP_808450.2 | 23 | 2832 | ||
GenBank mRNA | AK173168.1 | - | 23 | 2832 | ||
BC067047.1 | - | 21 | 2735 | |||
BC151074.1 | - | 23 | 2780 | 9087987 | ||
BC157940.1 | - | 23 | 2780 | 9088120 |