Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Commd8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178599.4 | NP_848714.1 | 3 | 342 | ||
GenBank mRNA | AK009275.1 | - | 3 | 331 | ||
AK012653.1 | - | 3 | 328 | |||
AK013015.1 | - | 4 | 413 | |||
AK013371.1 | - | 3 | 315 | |||
AK017777.1 | - | 4 | 356 | |||
AK077934.1 | - | 3 | 342 | |||
AK151560.1 | - | 3 | 325 | |||
AK158078.1 | - | 4 | 425 | |||
AK170011.1 | - | 3 | 315 | |||
BC051517.1 | - | 1 | 102 | |||
BC098183.1 | - | 3 | 318 | 5150466 |