Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cherp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138585.3 | NP_613051.3 | 16 | 2672 | ||
GenBank mRNA | AF536541.1 | - | 16 | 2568 | ||
AK005253.1 | - | 3 | 245 | |||
AK049765.1 | - | 16 | 2583 | |||
AK144328.1 | - | 16 | 2598 | |||
AK171907.1 | - | 9 | 1642 | |||
AK207608.1 | - | 2 | 43 | |||
BC032191.1 | - | 16 | 2584 | 5322208 |