Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Reep3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204915.1 | NP_001191844.1 | 4 | 376 | ||
NM_178606.5 | NP_848721.1 | 4 | 376 | |||
GenBank mRNA | AK005026.1 | - | 4 | 324 | ||
AK008058.1 | - | 4 | 295 | |||
AK012403.1 | - | 4 | 320 | |||
AK028868.1 | - | 4 | 333 | |||
AK047728.1 | - | 4 | 244 | |||
AK079649.1 | - | 4 | 246 | |||
AK154559.1 | - | 4 | 234 | |||
AK161684.1 | - | 1 | 1345 | |||
AY562231.1 | - | 4 | 209 | |||
BC004607.1 | - | 4 | 285 | 3585133 | ||
CJ163373.1 | - | 4 | 378 |