Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Serpinb1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173052.2 | NP_766640.1 | 5 | 578 | ||
GenBank mRNA | AF426025.1 | - | 4 | 461 | ||
AK018226.1 | - | 5 | 557 | |||
BC030354.1 | - | 5 | 523 | 5367365 |