Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mga | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164274.1 | NP_001157746.1 | 3 | 1463 | ||
NM_013720.2 | NP_038748.2 | 3 | 1463 | |||
GenBank mRNA | AF205935.1 | - | 2 | 1277 | ||
AK032570.1 | - | 3 | 1444 | |||
AK046252.1 | - | 3 | 1464 | |||
AK132911.1 | - | 1 | 1740 | |||
AK133775.1 | - | 3 | 1680 | |||
AK139919.1 | - | 3 | 1415 | |||
AK145491.1 | - | 3 | 1412 | |||
AK162302.1 | - | 3 | 1460 | |||
AK172962.1 | - | 3 | 1386 |