Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mga | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164274.1 | NP_001157746.1 | 8 | 3227 | ||
NM_013720.2 | NP_038748.2 | 8 | 3227 | |||
GenBank mRNA | AF205935.1 | - | 7 | 3041 | ||
AK032570.1 | - | 6 | 2971 | |||
AK046252.1 | - | 8 | 3228 | |||
AK139919.1 | - | 8 | 3180 | |||
AK172962.1 | - | 8 | 3150 |