Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hip1r | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145070.3 | NP_659507.3 | 3 | 306 | ||
GenBank mRNA | AF221713.1 | - | 3 | 171 | ||
AK014542.1 | - | 3 | 284 | |||
AK129182.1 | - | 3 | 262 | |||
AK146663.1 | - | 3 | 187 | |||
BC062914.1 | - | 3 | 187 | |||
BC065101.1 | - | 3 | 295 | 30535741 | ||
CJ082429.1 | - | 3 | 307 |