Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pde7b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013875.5 | NP_038903.3 | 6, 7 | 852 | ||
GenBank mRNA | AF190639.1 | - | 6, 7 | 633 | ||
AJ251859.1 | - | 6, 7 | 904 | |||
AK035385.1 | - | 6, 7 | 864 | |||
AK049910.1 | - | 6, 7 | 908 | |||
BC130267.1 | - | 6, 7 | 733 | 40143060 |