Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Iqgap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016721.2 | NP_057930.2 | 29 | 3856 | ||
GenBank mRNA | AF240630.1 | - | 29 | 3819 | ||
AK029631.1 | - | 11 | 1553 | |||
AK077354.1 | - | 5 | 781 | |||
AK129044.1 | - | 29 | 3794 | |||
AK154213.1 | - | 4 | 571 | |||
AK160892.1 | - | 11 | 1391 | |||
AK184833.1 | - | 2 | 210 | |||
BC046385.1 | - | 29 | 3774 | 3256630 |