Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdlim4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019417.3 | NP_062290.2 | 2 | 298 | ||
GenBank mRNA | AB221672.1 | - | 2 | 213 | ||
AK035513.1 | - | 2 | 273 | |||
BC030068.1 | - | 2 | 227 | 5144166 | ||
Y08361.1 | - | 2 | 249 |