Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kdm2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013910.2 | NP_038938.1 | 6 | 1009 | ||
GenBank mRNA | AK031626.1 | - | 6 | 975 | ||
AK129479.1 | - | 17 | 2446 | |||
BC057622.1 | - | 6 | 1004 | 6402440 | ||
BC065090.1 | - | 17 | 2645 | 6851421 | ||
BC068239.1 | - | 14 | 2199 |