Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mlf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170341.1 | NP_001163812.1 | 7, 8 | 786 | ||
NM_145385.2 | NP_663360.1 | 7, 8 | 786 | |||
GenBank mRNA | AK143972.1 | - | 7, 8 | 743 | ||
BC003975.1 | - | 7, 8 | 697 | 3488851 |