Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mlf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170341.1 | NP_001163812.1 | 3 | 323 | ||
NM_145385.2 | NP_663360.1 | 3 | 323 | |||
GenBank mRNA | AK141241.1 | - | 3 | 279 | ||
AK143972.1 | - | 3 | 280 | |||
AK171220.1 | - | 371 | ||||
BC003975.1 | - | 3 | 234 | 3488851 |