Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pttg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001131054.1 | NP_001124526.1 | 3 | 174 | ||
NM_013917.2 | NP_038945.2 | 2 | 426 | |||
GenBank mRNA | AF069051.1 | - | 2 | 418 | ||
AF071209.1 | - | 2 | 426 | |||
AK002473.1 | - | 2 | 140 | |||
AK008704.1 | - | 2 | 141 | |||
AK088387.1 | - | 2 | 140 | |||
BC023324.1 | - | 3 | 180 | 4946234 | ||
CN837307.1 | - | 3 | 179 | |||
DV657646.1 | - | 2 | 144 |