Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013919.4 | NP_038947.2 | 2 | 264 | ||
GenBank mRNA | AF177759.1 | - | 1 | 63 | ||
AK011148.1 | - | 2 | 265 | |||
AK013252.1 | - | 2 | 259 | |||
AK049649.1 | - | 1 | 45 | |||
BC021903.1 | - | 2 | 254 | 3985792 | ||
BY110597.1 | - | 2 | 264 | |||
KF646669.1 | - | 1 | 43 |