Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bin1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083334.1 | NP_001076803.1 | 4 | 483 | ||
NM_009668.2 | NP_033798.1 | 4 | 483 | |||
GenBank mRNA | BC065160.1 | - | 4 | 455 | 5718309 | |
BQ573238.1 | - | 4 | 454 | |||
BU707946.1 | - | 3 | 185 | |||
U60884.1 | - | 4 | 471 |