Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lcmt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025304.3 | NP_079580.2 | 5, 6 | 616 | ||
GenBank mRNA | AK003424.1 | - | 5, 6 | 541 | ||
BC132507.1 | - | 5, 6 | 522 | 40130784 | ||
BC132509.1 | - | 5, 6 | 522 | 40130786 | ||
BC145301.1 | - | 5, 6 | 522 |