Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mapk8ip3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163447.1 | NP_001156919.1 | 19 | 2290 | ||
NM_001163448.1 | NP_001156920.1 | 18 | 2248 | |||
NM_001163449.1 | NP_001156921.1 | 20 | 2224 | |||
NM_001163450.1 | NP_001156922.1 | 19 | 2200 | |||
NM_001163451.1 | NP_001156923.1 | 19 | 2197 | |||
NM_001163453.1 | NP_001156925.1 | 18 | 2182 | |||
NM_013931.4 | NP_038959.2 | 19 | 2293 | |||
GenBank mRNA | AB005662.1 | - | 19 | 2180 | ||
AB043123.1 | - | 20 | 2101 | |||
AB043124.1 | - | 19 | 2170 | |||
AB043125.1 | - | 19 | 2167 | |||
AB093282.2 | - | 19 | 2152 | |||
AF178636.1 | - | 18 | 2165 | |||
AF178637.1 | - | 19 | 2286 | |||
AF262046.1 | - | 19 | 2276 | |||
BC060603.1 | - | 20 | 2159 | 5706805 | ||
BC065105.1 | - | 18 | 2172 | 30356817 | ||
BC068312.1 | - | 19 | 2271 | 30100914 | ||
CO433928.1 | - | 1 | 30 |