Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vapa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013933.3 | NP_038961.2 | 6 | 911 | ||
GenBank mRNA | AF115503.1 | - | 6 | 633 | ||
AF157497.1 | - | 6 | 852 | |||
AK003576.1 | - | 6 | 852 | |||
AK012357.1 | - | 6 | 666 | |||
AK088132.1 | - | 6 | 888 | |||
AK149040.1 | - | 518 | ||||
AK159582.1 | - | 6 | 853 | |||
AK167381.1 | - | 6 | 912 | |||
AK168824.1 | - | 6 | 858 | |||
BC003866.1 | - | 6 | 897 | 3490082 |