Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankef1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175667.4 | NP_783598.1 | 7 | 1808 | ||
GenBank mRNA | AK029804.1 | - | 1944 | |||
BC089504.1 | - | 7 | 1849 | |||
BC120825.1 | - | 7 | 1808 | 40129748 | ||
BC144833.1 | - | 7 | 1823 |