Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcmtd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183028.3 | NP_898849.1 | 3 | 767 | ||
GenBank mRNA | AK040848.1 | - | 3 | 495 | ||
AK079468.1 | - | 2 | 304 | |||
AK145008.1 | - | 3 | 493 | |||
AK166166.1 | - | 3 | 652 | |||
AK166593.1 | - | 3 | 493 | |||
AK168606.1 | - | 3 | 519 | |||
AK168705.1 | - | 3 | 537 | |||
AK187882.1 | - | 2 | 76 | |||
BC019193.1 | - | 3 | 515 | |||
BC052741.1 | - | 2 | 484 | 6833458 | ||
BC064801.1 | - | 2 | 364 | 30067935 | ||
BC110360.1 | - | 3 | 537 | 5326388 | ||
BC117941.1 | - | 2 | 484 | 40097836 | ||
BC117942.1 | - | 2 | 484 | 40097839 |