Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcmtd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183028.3 | NP_898849.1 | 6 | 1446 | ||
GenBank mRNA | AK079468.1 | - | 5 | 983 | ||
AK134223.1 | - | 2 | 379 | |||
AK136768.1 | - | 2 | 443 | |||
AK145008.1 | - | 6 | 1173 | |||
AK161921.1 | - | 3 | 1431 | |||
AK166166.1 | - | 6 | 1332 | |||
AK166593.1 | - | 6 | 1172 | |||
AK168606.1 | - | 6 | 1198 | |||
AK168705.1 | - | 6 | 1216 | |||
BC019193.1 | - | 6 | 1194 | |||
BC052741.1 | - | 5 | 1163 | 6833458 | ||
BC064801.1 | - | 5 | 1043 | 30067935 | ||
BC110360.1 | - | 6 | 1216 | 5326388 | ||
BC117941.1 | - | 5 | 1163 | 40097836 | ||
BC117942.1 | - | 5 | 1163 | 40097839 |