Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fndc3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207636.2 | NP_997519.2 | 21, 22 | 2806 | ||
GenBank mRNA | AK048586.1 | - | 21, 22 | 2809 | ||
AK089380.1 | - | 21, 22 | 2771 | |||
AK129250.1 | - | 7, 8 | 753 | |||
AK154790.1 | - | 21, 22 | 2575 | |||
BC047066.1 | - | 5, 6 | 570 | |||
BC094678.1 | - | 12, 13 | 1445 | |||
BC144880.1 | - | 21, 22 | 2614 | |||
DQ192036.1 | - | 21, 22 | 2807 |