Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppm1h | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110218.1 | NP_001103688.1 | 4 | 1202 | ||
NM_176919.4 | NP_795893.2 | 4 | 1202 | |||
GenBank mRNA | AK029461.1 | - | 4 | 764 | ||
AK040194.1 | - | 4 | 946 | |||
AK040207.1 | - | 4 | 1187 | |||
AK043121.1 | - | 4 | 1203 | |||
AK051434.1 | - | 4 | 1161 | |||
AK077422.1 | - | 4 | 1198 | |||
AK134804.1 | - | 4 | 1182 | |||
AK220260.1 | - | 4 | 1159 | |||
BC052910.1 | - | 4 | 513 |