Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr59 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170742.1 | NP_001164213.1 | 18, 19 | 2001 | ||
NM_176923.4 | NP_795897.2 | 18, 19 | 2001 | |||
GenBank mRNA | AK030644.1 | - | 18, 19 | 1882 | ||
AK040807.1 | - | 3, 4 | 288 | |||
AK136303.1 | - | 14, 15 | 1553 | |||
AK170910.1 | - | 18, 19 | 2000 | |||
BC066082.1 | - | 18, 19 | 1986 | 30355918 | ||
EF011622.1 | - | 18, 19 | 1986 |