Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nrcam | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146031.1 | NP_001139503.1 | 8 | 1176 | ||
NM_176930.4 | NP_795904.3 | 8 | 1176 | |||
GenBank mRNA | AJ543321.1 | - | 8 | 1078 | ||
AK045259.1 | - | 7 | 877 | |||
AK048567.1 | - | 9 | 1166 | |||
AK087693.1 | - | 8 | 1128 | |||
AK122252.1 | - | 8 | 1106 | |||
AK178439.1 | - | 1 | 76 | |||
AK214713.1 | - | 1 | 76 | |||
AK218057.1 | - | 3 | 255 | |||
BC055053.1 | - | 7 | 791 |
Set Membership: |
G-protein coupled receptor |