Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nrcam | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146031.1 | NP_001139503.1 | 7 | 974 | ||
NM_176930.4 | NP_795904.3 | 7 | 974 | |||
GenBank mRNA | AJ543321.1 | - | 7 | 876 | ||
AK045259.1 | - | 6 | 675 | |||
AK048567.1 | - | 8 | 964 | |||
AK087693.1 | - | 7 | 926 | |||
AK122252.1 | - | 7 | 904 | |||
AK218057.1 | - | 2 | 53 | |||
BC055053.1 | - | 6 | 589 |
Set Membership: |
G-protein coupled receptor |