Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Efcab5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_176965.3 | NP_795939.2 | 10 | 2542 | ||
GenBank mRNA | AK029790.1 | - | 8 | 2129 | ||
AK035698.1 | - | 6 | 1376 | |||
BC127154.1 | - | 8 | 2084 |