Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nt5dc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_176968.4 | NP_795942.2 | 9 | 987 | ||
GenBank mRNA | AK049656.1 | - | 8 | 902 | ||
AK077876.1 | - | 9 | 988 | |||
AK145666.1 | - | 9 | 988 | |||
AK164144.1 | - | 9 | 928 | |||
AK171761.1 | - | 9 | 997 | |||
BC098476.1 | - | 9 | 947 | 30051251 | ||
BC125276.1 | - | 9 | 986 | 40107511 |