Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cep295 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_176976.4 | NP_795950.2 | 15 | 4651 | ||
GenBank mRNA | AK030838.1 | - | 1 | 948 | ||
AK032664.1 | - | 1 | 578 | |||
BC085189.1 | - | 1 | 840 | |||
BC118968.2 | - | 1 | 713 |