Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxo48 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_176982.2 | NP_795956.1 | 3 | 1285 | ||
GenBank mRNA | AK037835.1 | - | 3 | 1285 | ||
AK041978.1 | - | 3 | 592 | |||
BC125589.1 | - | 2 | 162 | 40130104 | ||
BC125591.1 | - | 2 | 162 | 40130106 | ||
BC145147.1 | - | 1 | 257 |