Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bbs9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178415.1 | NP_848502.1 | 7 | 864 | ||
NM_181316.4 | NP_851833.2 | 6 | 808 | |||
GenBank mRNA | AK028200.1 | - | 6 | 898 | ||
AK086114.1 | - | 6 | 780 | |||
AK144730.1 | - | 7 | 835 | |||
AK166297.1 | - | 6 | 786 | |||
BC030419.1 | - | 3 | 157 | |||
BC046426.1 | - | 6 | 820 | 3601505 |