Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bbs9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178415.1 | NP_848502.1 | 3 | 473 | ||
NM_181316.4 | NP_851833.2 | 2 | 417 | |||
GenBank mRNA | AK028200.1 | - | 2 | 507 | ||
AK086114.1 | - | 2 | 388 | |||
AK144730.1 | - | 3 | 444 | |||
AK166297.1 | - | 2 | 395 | |||
BC046426.1 | - | 2 | 429 | 3601505 |