Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Apba1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177034.3 | NP_796008.2 | 5, 6 | 1785 | ||
GenBank mRNA | AK032261.1 | - | 4, 5 | 549 | ||
AK147583.1 | - | 5, 6 | 1775 | |||
AK188637.1 | - | 1, 2 | 139 | |||
BC141181.1 | - | 5, 6 | 1603 | 9055793 |