Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Casc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001205369.1 | NP_001192298.1 | 1 | 433 | ||
NM_001205370.1 | NP_001192299.1 | 1 | 433 | |||
NM_177054.5 | NP_796028.2 | 1 | 433 | |||
NM_199038.3 | NP_950239.2 | 1 | 433 | |||
GenBank mRNA | AK051613.1 | - | 1 | 275 | ||
AK080364.1 | - | 1 | 284 | |||
AK160708.1 | - | 1 | 274 | |||
AY484583.1 | - | 1 | 273 | |||
AY484585.1 | - | 1 | 273 | |||
BC034556.1 | - | 1 | 296 | |||
BC064446.1 | - | 1 | 433 | 6843743 |