Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Exph5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_176846.3 | NP_789816.2 | 6 | 4641 | ||
GenBank mRNA | AB098163.1 | - | 6 | 4641 | ||
AK143980.1 | - | 1 | 2336 | |||
AK172984.1 | - | 1 | 3567 | |||
BC120905.1 | - | 6 | 4628 | 40087301 |