Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B630019A10Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK046764.1 | - | 3 | 2305 |
Gene Symbol: BC106179 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK041922.1 | - | 2 | 114 | ||
AK161755.1 | - | 2 | 399 | |||
BC106179.1 | - | 2 | 395 | 6509459 |