Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cep95 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166685.1 | NP_001160157.1 | 8 | 971 | ||
NM_177088.3 | NP_796062.2 | 9 | 1100 | |||
GenBank mRNA | AK052705.1 | - | 9 | 1028 | ||
AK076400.1 | - | 9 | 1078 | |||
BC090675.1 | - | 9 | 1056 | |||
BC116308.1 | - | 8 | 937 | 40095242 | ||
BC116309.1 | - | 8 | 950 |