Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tacc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177089.5 | NP_796063.3 | 8 | 2032 | ||
NM_199323.3 | NP_955355.1 | 6 | 808 | |||
GenBank mRNA | AK158441.1 | - | 8 | 2039 | ||
AY177412.1 | - | 6 | 716 | |||
AY177413.1 | - | 8 | 1934 | |||
BC125389.1 | - | 6 | 618 | |||
BC145709.1 | - | 6 | 618 |