Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Senp5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177103.4 | NP_796077.2 | 4, 5 | 2061 | ||
GenBank mRNA | AK028177.1 | - | 4, 5 | 2064 | ||
AK042087.1 | - | 4, 5 | 1954 | |||
AK043171.1 | - | 4, 5 | 1946 | |||
AK136120.1 | - | 3, 4 | 1654 | |||
AK158338.1 | - | 4, 5 | 1927 | |||
BC067014.1 | - | 4, 5 | 1828 | 6812169 | ||
BC080830.1 | - | 4, 5 | 1945 | 30433573 |