Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttll5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081423.2 | NP_001074892.2 | 22 | 2312 | ||
GenBank mRNA | AB093278.1 | - | 22 | 2245 | ||
AK009255.1 | - | 6 | 690 | |||
AK031470.1 | - | 3 | 384 | |||
AM690748.1 | - | 21 | 2077 | |||
BC035276.1 | - | 21 | 2129 |