Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fryl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028194.2 | NP_082470.2 | 21 | 2735 | ||
GenBank mRNA | AK036035.1 | - | 19 | 2453 | ||
AK164590.1 | - | 21 | 2737 | |||
BQ930071.1 | - | 2 | 205 |