Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cenpt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177150.2 | NP_796124.1 | 3 | 296 | ||
GenBank mRNA | AK089172.1 | - | 3 | 343 | ||
AK090340.1 | - | 3 | 297 | |||
AK143613.1 | - | 3 | 259 | |||
AK167376.1 | - | 3 | 297 | |||
AK182753.1 | - | 2 | 295 | |||
BC080832.1 | - | 2 | 273 | |||
BC100321.1 | - | 2 | 399 | |||
BC121824.1 | - | 3 | 297 | 40107149 |