Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: A830018L16Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160369.2 | NP_001153841.1 | 9, 10 | 1522 | ||
NM_177173.6 | NP_796147.2 | 9, 10 | 1513 | |||
GenBank mRNA | AK034391.1 | - | 9, 10 | 1509 | ||
AK081622.1 | - | 9, 10 | 1545 | |||
BC137903.1 | - | 9, 10 | 1118 | 8860922 | ||
BC145022.1 | - | 9, 10 | 1118 | 9053570 | ||
BC145023.1 | - | 9, 10 | 1118 | 9053571 |