Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zbtb26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199025.2 | NP_950190.2 | 2 | 1656 | ||
GenBank mRNA | AK044592.1 | - | 2 | 1287 | ||
AK173205.1 | - | 2 | 1281 | |||
BC060655.1 | - | 2 | 1656 | 30355035 | ||
BC109349.1 | - | 2 | 1639 | 40056733 | ||
BC109350.1 | - | 2 | 1639 | 40056735 |