Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Enpp6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177304.3 | NP_796278.1 | 6 | 1007 | ||
GenBank mRNA | AK014741.1 | - | 1 | 63 | ||
AK038460.1 | - | 6 | 979 | |||
AK046797.1 | - | 6 | 1008 | |||
AK046881.1 | - | 6 | 1012 | |||
AK079352.1 | - | 5 | 1349 | |||
AK079642.1 | - | 6 | 1007 | |||
BC096376.1 | - | 6 | 978 | 4237600 |