Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rfx6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159389.1 | NP_001152861.1 | 17 | 2281 | ||
NM_177306.3 | NP_796280.1 | 14 | 1912 | |||
GenBank mRNA | AK029809.1 | - | 14 | 1913 | ||
BC132104.1 | - | 12 | 1595 | 40130381 | ||
BC132294.1 | - | 12 | 1595 | 40130571 | ||
BC145065.1 | - | 11 | 1477 | |||
FJ804414.1 | - | 16 | 2129 |