Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpr183 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183031.2 | NP_898852.2 | 2 | 1035 | ||
GenBank mRNA | AK087951.1 | - | 2 | 947 | ||
AK138693.1 | - | 2 | 1037 | |||
AK141593.1 | - | 2 | 948 | |||
AK153216.1 | - | 2 | 948 | |||
AK156005.1 | - | 2 | 948 | |||
AK156289.1 | - | 2 | 955 | |||
AY255606.1 | - | 1 | 123 | |||
BC052868.1 | - | 2 | 942 | 30073287 |
Set Membership: |
G-protein coupled receptor |